Skip to Content
Merck
All Photos(1)

Key Documents

870715P

Avanti

15:0 Coenzyme A

Avanti Research - A Croda Brand 870715P, powder

Synonym(s):

pentadecanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H73N10O17P3S
CAS Number:
Molecular Weight:
1043.01
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870715P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870715P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

General description

15:0 Coenzyme A, also known as pentadecanoyl Coenzyme A, is a coenzyme A derivative of pentadecanoic acid. 15:0 Coenzyme A is a long-chain acyl CoA and the product of the first round of β-oxidation. It is a substrate for serine palmitoyltransferase (SPT) for de novo synthesis of C17-long chain bases.

Application

15:0 Coenzyme A has been used as a constituent of reaction cocktail in microsomal serine palmitoyltransferase (SPT) assay.

Packaging

5 mL Amber Glass Screw Cap Vial (870715P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of isotopically labeled substrates reveals kinetic differences between human and bacterial serine palmitoyltransferase
Harrison PJ, et al.
Journal of Lipid Research, 60(5), 953-962 (2019)
Analysis of coenzyme A activated compounds in actinomycetes
Cabruja M, et al.
Applied Microbiology and Biotechnology, 100(16), 7239-7248 (2016)
Peter J Harrison et al.
Journal of lipid research, 60(5), 953-962 (2019-02-23)
Isotope labels are frequently used tools to track metabolites through complex biochemical pathways and to discern the mechanisms of enzyme-catalyzed reactions. Isotopically labeled l-serine is often used to monitor the activity of the first enzyme in sphingolipid biosynthesis, serine palmitoyltransferase
Matías Cabruja et al.
Applied microbiology and biotechnology, 100(16), 7239-7248 (2016-06-09)
Acyl-CoAs are crucial compounds involved in essential metabolic pathways such as the Krebs cycle and lipid, carbohydrate, and amino acid metabolisms, and they are also key signal molecules involved in the transcriptional regulation of lipid biosynthesis in many organisms. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service