Skip to Content
Merck
All Photos(2)

Key Documents

860103C

Avanti

18:1 Phosphatidylpropanol

1,2-dioleoyl-sn-glycero-3-phosphopropanol (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphopropanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78O8PNa
CAS Number:
Molecular Weight:
765.03
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860103C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860103C

concentration

10 mg/mL (860103C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC)OCCC

InChI

1S/C42H79O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41(43)47-38-40(39-49-51(45,46)48-37-6-3)50-42(44)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h19-22,40H,4-18,23-39H2,1-3H3,(H,45,46);/q;+1/p-1/b21-19-,22-20-;/t40-;/m1./s1

InChI key

ZGATVCGNENIEOO-MYJLHYLNSA-M

Application

18:1 Phosphatidylpropanol or 1,2-dioleoyl-sn-glycero-3-phosphopropanol has been used in the preparation of doped giant unilamellar vesicles (GUVs). It is suitable for use as an internal standard in ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) to evaluate its matrix effect in blood samples for the validation of phosphatidylethanol (PEth). It may also be used as an internal standard for the quantification of PEth using liquid chromatography tandem mass spectrometry (LC-MS/MS) in human blood.

Packaging

5 mL Clear Glass Sealed Ampule (860103C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlling loading and optical properties of gold nanoparticles on liposome membranes
Sau TK, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 342(1-3), 92-96 (2009)
Xinyu Zhang et al.
Analytica chimica acta, 963, 68-75 (2017-03-25)
The importance of ethanol non-oxidative metabolites as the specific biomarkers of alcohol consumption in clinical and forensic settings is increasingly acknowledged. Simultaneous determination of these metabolites can provide a wealth of information like drinking habit and history, but it was
Natalie Kummer et al.
Analytical and bioanalytical chemistry, 408(3), 825-838 (2015-11-26)
Phosphatidylethanol species (PEths) are promising biomarkers of alcohol consumption. Here, we report on the set-up, validation, and application of a novel UHPLC-ESI-MS/MS method for the quantification of PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 in whole blood (30 μL) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service