Skip to Content
Merck
All Photos(1)

Key Documents

810290P

Avanti

18:2 TopFluor cholesterol

Avanti Research - A Croda Brand 810290P, powder

Synonym(s):

23-(dipyrrometheneboron difluoride)-24-norcholesteryl linoleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H81 BF2N2O2
CAS Number:
Molecular Weight:
839.04
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810290P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810290P

shipped in

dry ice

storage temp.

−20°C

General description

18:2 TopFluor cholesterol or 23-(dipyrrometheneboron difluoride)-24-norcholesteryl linoleate is a cholesterol analog containing unsaturated (18:2) acyl chain.

Application

18:2 TopFluor cholesterol or 23-(dipyrrometheneboron difluoride)-24-norcholesteryl linoleate might be used to study the diffusion of cholesterol analogs compared with phospholipid analogs in model membranes.

Packaging

5 mL Amber Glass Screw Cap Vial (810290P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Pinkwart et al.
The Journal of biological chemistry, 294(34), 12599-12609 (2019-07-05)
Cholesterol constitutes ∼30-40% of the mammalian plasma membrane, a larger fraction than of any other single component. It is a major player in numerous signaling processes as well as in shaping molecular membrane architecture. However, our knowledge of the dynamics
Xiandeng Wu et al.
Molecular cell, 73(5), 971-984 (2019-01-22)
Both the timing and kinetics of neurotransmitter release depend on the positioning of clustered Ca2+ channels in active zones to docked synaptic vesicles on presynaptic plasma membranes. However, how active zones form is not known. Here, we show that RIM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service