Z699470
Dilution bottle
Butterfield′s Buffer, volume 90 mL
Synonym(s):
dilution bottle, dilution bottles, phosphate buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
Recommended Products
packaging
pkg of 72 ea
manufacturer/tradename
Maine Mfg 10498504
capacity
90 mL
volume
90 mL
Looking for similar products? Visit Product Comparison Guide
General description
Butterfield′s Phosphate Buffer contains monobasic potassium phosphate and is used extensively in the food, dairy and pharmaceutical industries. Offered in 90 mL and 99 mL volumes for easy 1:10 and 1:100 dilutions. It is recommended as a general diluent in laboratory procedures by the Federal Drug Administration and in the Bacteriological Analytical Manual. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing.
Whatman sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH ± 0.2 pH at 25°C. The buffers are supplied in an easy open, flip-top, plastic container with a tamper-evident seal.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service