Skip to Content
Merck
All Photos(1)

Key Documents

Z685682

Tygon® formula 2375 laboratory tubing

I.D. × O.D. 9.6 mm × 15.9 mm, L 15 m (50 ft)

Synonym(s):

chemical resistant tubing, laboratory tubing, non-leaching tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105108
NACRES:
NB.35

packaging

pack of 50 feet

manufacturer/tradename

St. Gobain AJK00029

I.D. × O.D.

9.6 mm × 15.9 mm

L

15 m (50 ft)

wall thickness

3.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

Tygon laboratory tubing, formula 2375, is chemically resistant tubing. Clear, flexible tubing suitable for specialty chemical production and hazardous material handling.

  • Resistant to MEK, acids, bases, ketones, and alcohols
  • Plasticizer free
  • No contamination from leaching
  • Biocompatible
  • No disposal concerns
  • Releases only carbon dioxide and water upon incineration
  • Sterilize by gas or radiation

Specifications

Technical data:
Tensile Strength: 1900 psi (13.1 MPa)
Ultimate Elongation: 850%
Tensile Stress @ 100% elongation: 425 psi (2.9 MPa)
Tensile Set (@ 75% of ultimate elongation): 300%
Tear Resistance: 240 lb-f/inch (42 kN/m)
Compression Set Constant Deflection: 100% @ 158°F (70°C) for 22 hours
Brittle Temperature: < -75°F (<-59°C)
Low Temp. Flexibility: -103°F (-75°C)
Specific Gravity: 0.9
Water Absorption: 0.04% 24 hrs at 73°F (23°C)
Maximum Recommended Operating Temperature: 130°F
Color: Clear

Legal Information

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service