Key Documents
Z555967
Scienceware® Touch Free automatic waste can
stainless steel, capacity 12 gal
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
stainless steel
manufacturer/tradename
Bel-Art
capacity
12 gal
General description
Opens and closes automatically, protecting users from contact with the waste receptacle or its contents when disposing of waste of any kind-biological/biohazard materials, paper trash, or recyclables. When a hand or solid object is moved over the top, the Scienceware Touch Free automatic waste can detects the motion and opens the lid to accept waste. It then closes automatically.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Touch Free is a trademark of SP Scienceware, a division of SP