WHA77040001
Whatman® UNISEAL microplate seals
thin cold sealing film, adhesive backing, clear polypropylene
Synonym(s):
Z746827
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
material
clear polypropylene
sterility
non-sterile
feature
thin cold sealing film, adhesive backing
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 7704-0001
Whatman Article No. 28420886 (US reference)
Related Categories
General description
Seals are used to control humidity and reduce evaporation of samples.
Features and Benefits
- Prevent spills and contamination.
- Cold seals are self-sticking with inert adhesive.
- Heat seals are available in a clear polyester or aluminum foil.
- Heat seals are for polypropylene microplates only and are applied with heat and pressure.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)