Skip to Content
Merck
All Photos(1)

Key Documents

W356700

Sigma-Aldrich

trans-p-Methoxycinnamaldehyde

≥98%, stabilized

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCHO
CAS Number:
Molecular Weight:
162.19
FEMA Number:
3567
Council of Europe no.:
11919
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

Assay

≥98%

contains

α-Tocopherol, synthetic as stabilizer

bp

145 °C/7 mmHg (lit.)

mp

55-60 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cinnamon; spicy

SMILES string

[H]C(=O)C(\[H])=C(/[H])c1ccc(OC)cc1

InChI

1S/C10H10O2/c1-12-10-6-4-9(5-7-10)3-2-8-11/h2-8H,1H3/b3-2+

InChI key

AXCXHFKZHDEKTP-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-p-Methoxycinnamaldehyde is an aromatic aldehyde reported to be found in Korean mint.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Five hTRPA1 agonists found in indigenous Korean mint, agastache rugosa.
Moon H, et al.
PLoS ONE, 10(5), e0127060-e0127060 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service