Skip to Content
Merck
All Photos(1)

Key Documents

W317004

Sigma-Aldrich

trans-3-Hexenoic acid

≥98%, FG

Synonym(s):

(E)-3-Hexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CO2H
CAS Number:
Molecular Weight:
114.14
FEMA Number:
3170
EC Number:
Council of Europe no.:
2256
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
08.050
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥98%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.44 (lit.)

bp

118-119 °C/22 mmHg (lit.)

mp

11-12 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cheese

SMILES string

CC\C=C\CC(O)=O

InChI

1S/C6H10O2/c1-2-3-4-5-6(7)8/h3-4H,2,5H2,1H3,(H,7,8)/b4-3+

InChI key

XXHDAWYDNSXJQM-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-3-Hexenoic acid occurs naturally in strawberry and bamboo leaves volatile oil.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition and antioxidant activity of the volatile oil of bamboo leaves from different provenances.
Liao HB, et al.
Applied Mechanics and Materials, 108, 320-325 (2012)
Christian Landmann et al.
Planta, 226(2), 417-428 (2007-02-27)
Fragaria x ananassa UDP-glucose:cinnamate glucosyltransferase (FaGT2) catalyzes the formation of cinnamic acid and p-coumaric acid glucose esters during strawberry fruit ripening. Here, the ripening and oxidative stress induced enzyme was further characterized by testing a range of structurally different substrates
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service