SYNA351024
Synthware™ distillation adapter
top joint: ST/NS 10/18, bottom joint: ST/NS 24/40, side joint: 24/40, immersion depth 50 mm, side arm angle: 75 deg
Synonym(s):
Synthware A351024, adapter, sidearm adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
bottom joint: ST/NS24/40
side joint: 24/40
joint: ST/NS top10/18
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A351024
immersion depth
50 mm
Related Categories
General description
For use as a component in atmospheric or vacuum distillations.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)