Skip to Content
Merck
All Photos(1)

Documents

P31000

Sigma-Aldrich

Phenylpropargyl aldehyde

96%

Synonym(s):

Phenylpropiolaldehyde, 3-Phenyl-2-propynal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCHO
CAS Number:
Molecular Weight:
130.14
Beilstein:
1099281
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.604 (lit.)

bp

118 °C/13 mmHg (lit.)

density

1.064 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)C#Cc1ccccc1

InChI

1S/C9H6O/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6,8H

InChI key

IDASOVSVRKONFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Grace et al.
The Journal of biological chemistry, 262(35), 16778-16785 (1987-12-15)
beta-Lactamases of all three classes, A, B, and C, are inactivated by phenylpropynal and p-nitrophenylpropynal. The inactivation of RTEM-2 beta-lactamase and of Bacillus cereus beta-lactamase I is accelerated in the presence of A type substrates such as dicloxacillin, quinacillin, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service