GF83071931
Polystyrene
(PS), film, thickness 0.125 mm, size 300 × 300 mm, clear clear
Synonym(s):
ST311125
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
film
manufacturer/tradename
Goodfellow 830-719-31
size
300 × 300 mm
thickness
0.125 mm
color
clear clear
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service