D45606
3-(Dibutylamino)propylamine
98%
Synonym(s):
N,N-Dibutyl-1,3-propanediamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[CH3(CH2)3]2N(CH2)3NH2
CAS Number:
Molecular Weight:
186.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.4463 (lit.)
bp
205 °C (lit.)
density
0.827 g/mL at 25 °C (lit.)
SMILES string
CCCCN(CCCC)CCCN
InChI
1S/C11H26N2/c1-3-5-9-13(10-6-4-2)11-7-8-12/h3-12H2,1-2H3
InChI key
KYCGURZGBKFEQB-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
219.2 °F - closed cup
Flash Point(C)
104 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Marieta Constantin et al.
International journal of biological macromolecules, 164, 4487-4498 (2020-09-19)
A new amphiphilic pullulan derivative (DBAP-PO) was obtained by grafting tertiary butyl amine and octanoyl groups on the pullulan backbone as cationic and hydrophobic moieties, respectively. The structural characteristics of the modified polymer were investigated by FT-IR and 1H and
Yi Wang et al.
Biomaterials, 112, 153-163 (2016-10-22)
Immunotherapy has shown promising treatment effects for a variety of cancers. However, the immune treatment efficiency for solid tumors is limited owing to insufficient infiltration of immune cells into solid tumors. The conversion of tumor-supportive macrophages to tumor-suppressive macrophages, inducing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service