Skip to Content
Merck
All Photos(1)

Key Documents

BR780775

BRAND® screw caps with tamper evident screw cap for micro tubes

yellow

Synonym(s):

Accessories, Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

polypropylene
yellow

packaging

pack of 1000 ea

manufacturer/tradename

BRAND 780775

Looking for similar products? Visit Product Comparison Guide

General description

Applications: -196 °C to +121 °C.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service