Key Documents
BR130324
BRAND® screw cap
thread for GL W 54
Synonym(s):
Accessories, Bottles, Screw caps
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
polypropylene
description
for wide neck bottles
packaging
pack of 1 ea
manufacturer/tradename
BRAND 130324
fitting
thread for GL W 54
General description
The BRAND® Screw Cap is a secure blue cap designed for square bottles, featuring a tamper-proof closure that ensures the integrity of the contents. This cap includes a press-off retaining ring and a sealing cone, providing an effective seal to prevent leaks and contamination. Its durable construction makes it suitable for various laboratory and industrial applications, ensuring reliable performance in sample storage and handling.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)