Skip to Content
Merck
All Photos(1)

Key Documents

AEM2A0522

Sigma-Aldrich

Xion AEM-Pention-72-5CL

greener alternative

thickness 5 μm, L × W 5 cm × 5 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104509
NACRES:
NB.21

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

L × W

5 cm × 5 cm

thickness

5 μm

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for use as Proton-Exchange Membranes (Fuel-Cell Membranes). Click here for more information.

Application

XION Composite PENTION Membranes are ultra-thin, ultra-strong, and provide ultra-high performance while demonstrating highly stable performance as reinforced anion exchange membranes (AEM). The ionomer structure contains a poly(norbornene) backbone with quaternary ammonium functional groups. A reinforcement layer is integrated into the structure of the membrane to provide enhanced mechanical properties. The enhanced mechanical properties as free-standing membranes, providing higher ionic conductance without sacrificing strength. Applications for XION Composite PENTION Membranes include fuel cells, electrolysis, and flow batteries.

Legal Information

Product of Xergy

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service