Skip to Content
Merck
All Photos(1)

Key Documents

930008

Sigma-Aldrich

RuCl(p-cymene)[(R,R)-Ts-DPEN]

Synonym(s):

[N-[(1R,2R)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35ClN2O2RuS
CAS Number:
Molecular Weight:
636.21
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

form

powder or crystals

Quality Level

mp

215 °C

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1.Cc2ccc(cc2)S(=O)(=O)N([Ru]Cl)[C@@H]([C@H](N)c3ccccc3)c4ccccc4

InChI

1S/C21H21N2O2S.C10H14.ClH.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-8(2)10-6-4-9(3)5-7-10;;/h2-15,20-21H,22H2,1H3;4-8H,1-3H3;1H;/q-1;;;+2/p-1/t20-,21-;;;/m1.../s1

InChI key

AZFNGPAYDKGCRB-AGEKDOICSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phosphine cationic ruthenium catalysts used for enantioselective hydrogenation of:
  • Quinolines
  • N-Alkyl ketimines
  • Antitumor and antiproliferative derivatives of natural products isolated from bacteria
  • Hydroxy arylaldehydes via Rap-Stoermer-enantioselective transfer hydrogenation

Catalyst with improved performance modified via microenvironment engineering of nanocages
RuCl(p-cymene)[(R,R)-Ts-DPEN] is a chiral diamine ligand complexed with ruthenium used for asymmetric hydrogenation reactions.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service