Skip to Content
Merck
All Photos(2)

Key Documents

904813

Sigma-Aldrich

(S,R,S)-AHPC-PEG2-azide

Synonym(s):

(2S,4R)-1-((S)-2-(2-(2-(2-Azidoethoxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, Crosslinker−E3 ligase ligand conjugate, Protein degrader building block for PROTAC® research, Template for synthesis of targeted protein degrader, VH032 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H39N7O6S
CAS Number:
Molecular Weight:
601.72
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

ligand

VH032

Assay

≥95%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: ligand-linker conjugate

functional group

azide

storage temp.

2-8°C

SMILES string

O=C(NCC1=CC=C(C2=C(C)N=CS2)C=C1)[C@H](C[C@@H](O)C3)N3C([C@H](C(C)(C)C)NC(COCCOCCN=[N+]=[N-])=O)=O

InChI

1S/C28H39N7O6S/c1-18-24(42-17-31-18)20-7-5-19(6-8-20)14-30-26(38)22-13-21(36)15-35(22)27(39)25(28(2,3)4)33-23(37)16-41-12-11-40-10-9-32-34-29/h5-8,17,21-22,25,36H,9-16H2,1-4H3,(H,30,38)(H,33,37)/t21-,22+,25-/m1/s1

InChI key

SDSUWMZOAFJCPF-OTNCWRBYSA-N

Application

Protein degrader builiding block (S,R,S)-AHPC-PEG2-azide enables the synthesis of molecules for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology. This conjugate contains a von Hippel-Lindau (VHL)-recruiting ligand and a PEGylated crosslinker with pendant azide for click chemistry with a target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and PROTAC, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a pendant azide group, parallel synthesis can be used to more quickly generate PROTAC libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Automate your VHL-PEG based PROTACs with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service