900276
MorDalPhos Pd G4
Synonym(s):
[4-[2-[Bis(tricyclo[3.3.1.13,7]dec-1-yl)phosphino-κP]phenyl]morpholine](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium
About This Item
form
powder
Quality Level
feature
generation 4
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings
functional group
phosphine
SMILES string
CNC1=C(C=CC=C1)C2=C(C=CC=C2)[Pd]OS(C)(=O)=O.C3(N4CCOCC4)=CC=CC=C3P([C@@]56CC(C[C@@H]7C6)CC(C7)C5)[C@@]89C[C@H](C[C@@H]%10C9)C[C@H](C%10)C8
InChI
1S/C30H42NOP.C13H12N.CH4O3S.Pd/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h1-4,21-26H,5-20H2;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1/t21-,22?,23?,24-,25?,26?,29-,30-,33?;;;
InChI key
VWMHZTMMTNRWLL-GYEYKPDQSA-M
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service