751634
Poly(ethylene glycol) methyl ether (4-cyano-4-pentanoate dodecyl trithiocarbonate)
average Mn 2,400
Synonym(s):
Dodecyl-trithiocarbonate-cyano PEG, PEG-RAFT, Poly(ethylene glycol)methyl ether 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentanoate
About This Item
Recommended Products
form
solid
mol wt
average Mn 2,400
transition temp
Tg 48-53 °C
PDI
≤1.1
Ω-end
4-cyano-4-pentanoate dodecyl trithiocarbonate
α-end
methoxy
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Molecular weight: PEG average Mn = 2,000; n~45
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service