Skip to Content
Merck
All Photos(3)

Key Documents

692980

Sigma-Aldrich

(S)-DTBM-SEGPHOS®

≥94%

Synonym(s):

(S)-(+)-5,5′-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′-bi-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H100O8P2
CAS Number:
Molecular Weight:
1179.53
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94%

form

powder

functional group

phosphine

SMILES string

COc1c(cc(cc1C(C)(C)C)P(c2cc(c(OC)c(c2)C(C)(C)C)C(C)(C)C)c3ccc4OCOc4c3-c5c6OCOc6ccc5P(c7cc(c(OC)c(c7)C(C)(C)C)C(C)(C)C)c8cc(c(OC)c(c8)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C74H100O8P2/c1-67(2,3)47-33-43(34-48(61(47)75-25)68(4,5)6)83(44-35-49(69(7,8)9)62(76-26)50(36-44)70(10,11)12)57-31-29-55-65(81-41-79-55)59(57)60-58(32-30-56-66(60)82-42-80-56)84(45-37-51(71(13,14)15)63(77-27)52(38-45)72(16,17)18)46-39-53(73(19,20)21)64(78-28)54(40-46)74(22,23)24/h29-40H,41-42H2,1-28H3

InChI key

ZNORAFJUESSLTM-UHFFFAOYSA-N

Application

Catalytic ligand for:
  • Enantioselective synthesis of secondary allylic alcohols via enantioselective reductions of α,β-unsaturated ketones
  • Stereoselective preparation of cyclohexanone derivatives via rhodium-catalyzed rearrangement of allyl or allenic cyclobutanols
  • Cu-catalyzed asymmetric conjugate reduction of beta-substituted unsaturated phosphonates to give optically active beta-stereogenic alkylphosphonates
  • Asymmetric synthesis of cyclohexenones via Rh-catalyzed desymmetrization through enantioselective C-C bond activation and ring expansion of allenyl cyclobutanols
  • Enantioselective synthesis and crystal structure of P-stereogenic alkynylphosphine oxides by Rh-catalyzed [2+2+2] cycloaddition

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service