Skip to Content
Merck
All Photos(1)

Key Documents

471267

Sigma-Aldrich

Dimethyl sulfoxide

≥99.6%, ReagentPlus®

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99.6%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

dilution

(for general lab use)

impurities

≤0.10% water
water

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Legal Information

Product of Arkema Inc.
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dongqing Li et al.
The Journal of clinical investigation, 125(8), 3008-3026 (2015-06-30)
Wound healing is a complex process that is characterized by an initial inflammatory phase followed by a proliferative phase. This transition is a critical regulatory point; however, the factors that mediate this process are not fully understood. Here, we evaluated
C Mathieu et al.
Journal of virology, 89(2), 1143-1155 (2014-11-08)
Measles virus (MV) infection is undergoing resurgence and remains one of the leading causes of death among young children worldwide despite the availability of an effective measles vaccine. MV infects its target cells by coordinated action of the MV H
Meng Wang et al.
Cancer research, 74(10), 2825-2834 (2014-03-22)
Therapeutics that target the epidermal growth factor receptor (EGFR) can enhance the cytotoxic effects of ionizing radiation (IR). However, predictive genomic biomarkers of this radiosensitization have remained elusive. By screening 40 non-small cell lung cancer cell (NSCLC) lines, we established
Mike Clements et al.
Toxicological sciences : an official journal of the Society of Toxicology, 140(2), 445-461 (2014-05-09)
Human stem cell derived cardiomyocytes (hESC-CM) provide a potential model for development of improved assays for pre-clinical predictive drug safety screening. We have used multi-electrode array (MEA) analysis of hESC-CM to generate multi-parameter data to profile drug impact on cardiomyocyte
D B Bas et al.
European journal of pain (London, England), 19(2), 260-270 (2014-06-20)
Mounting evidence points to individual contributions of tumour necrosis factor-alpha (TNF) and the c-Jun N-terminal kinase (JNK) pathway to the induction and maintenance of various pain states. Here we explore the role of spinal TNF and JNK in carrageenan-induced hypersensitivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service