406767
Poly[(phenyl glycidyl ether)-co-formaldehyde]
average Mn ~570
Synonym(s):
Phenol-formaldehyde polymer glycidyl ether, Phenol-formaldehyde polymer, oxiranylmethyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
specific activity
3.6 epoxide groups per molecule
mol wt
average Mn ~570
refractive index
n20/D 1.598
viscosity
200-500 poise(lit.)
density
1.227 g/mL at 25 °C (lit.)
SMILES string
[H]C([H])=O.C1OC1COc2ccccc2
Looking for similar products? Visit Product Comparison Guide
Application
Adhesives, structural and electrical laminates and coatings and castings for high temperature service.
Features and Benefits
Good chemical resistance at high temperatures. Substitutes for Bisphenol A and phenolic epoxy resins.
Physical form
Oligomeric epoxide.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service