Skip to Content
Merck
All Photos(2)

Key Documents

406309

Sigma-Aldrich

1-Chloro-4-propoxy-9H-thioxanthen-9-one

97%

Synonym(s):

1-Chloro-4-propoxy-9H -thioxanthone, 1-Chloro-4-propoxythioxanthone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClO2S
CAS Number:
Molecular Weight:
304.79
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

mp

99-103 °C (lit.)

SMILES string

CCCOc1ccc(Cl)c2C(=O)c3ccccc3Sc12

InChI

1S/C16H13ClO2S/c1-2-9-19-12-8-7-11(17)14-15(18)10-5-3-4-6-13(10)20-16(12)14/h3-8H,2,9H2,1H3

InChI key

VKQJCUYEEABXNK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel A Lago et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 33(3), 518-529 (2016-01-21)
Since the UV ink photoinitiator (PI) isopropylthioxanthone (ITX) was discovered in packaged milk, studies of print contamination have focused primarily on PIs but have also included amine synergists. Many other substances are used or formed during the print process, yet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service