Skip to Content
Merck
All Photos(1)

Key Documents

362573

Sigma-Aldrich

2-Chloro-6-methyl-3-pyridinecarbonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClN2
CAS Number:
Molecular Weight:
152.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

bp

140-145 °C (lit.)

mp

114-116 °C (lit.)

SMILES string

Cc1ccc(C#N)c(Cl)n1

InChI

1S/C7H5ClN2/c1-5-2-3-6(4-9)7(8)10-5/h2-3H,1H3

InChI key

YSBNBAYNISAUIT-UHFFFAOYSA-N

Application

2-Chloro-6-methyl-3-pyridinecarbonitrile (2-chloro-6-methylnicotinonitrile) may be used in the preparation of methyl-2-nitropyridinecarboxamides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Morisawa et al.
Journal of medicinal chemistry, 21(2), 194-199 (1978-02-01)
A number of methyl-2- and 3-nitropyridinecarboxamides have been synthesized. It has been established that the presence of at least one hydrogen atom, adjacent to the NO2 function, is important for anticoccidial activity and introduction of a methyl group to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service