Skip to Content
Merck
All Photos(1)

Key Documents

256544

Sigma-Aldrich

Cadmium telluride

powder, <250 μm, deposition grade, ≥99.98% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdTe
CAS Number:
Molecular Weight:
240.01
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

≥99.98% trace metals basis

form

powder

particle size

<250 μm

bp

1130 °C

mp

1092 °C

density

6.2 g/mL at 25 °C (lit.)

SMILES string

[Cd]=[Te]

InChI

1S/Cd.Te

InChI key

RPPBZEBXAAZZJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CdTe is a photonic material. Crystal structure of CdTe is zinc blende structure. 3CdTe is usually an obvious choice for photovoltaic applications because of its; low cost, direct band gap (1.44eV),3 high absorption coefficient, easily doped with p and n type dopants by several preparation techniques. 2

Application

Cadmium telluride may be used in photovoltaic solar cells as the window layer. CdTe was used in the preparation of an Au/Cu2Te/CdTe/CdS/TCO/glass heterostructure for solar cell.

Packaging

Packaged in poly bottles

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Au/Te/CdTe/CdS/TCO/Glass Solar Cells with Obtained by Sol-Gel as TCO
Marquez Marin J, et al.
International Journal of Photoenergy, 2014 (2014)
Ruizheng Liang et al.
Chemical communications (Cambridge, England), 49(10), 969-971 (2012-12-13)
Ordered ultrathin films based on alternate assembly of CdTe QDs and layered double hydroxide monolayers have been fabricated, which can be used as a photoluminescence temperature sensor with dual-parameter signals and high response sensitivity.
Asad Syed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 41-47 (2013-01-30)
The growing demand for semiconductor [quantum dots (Q-dots)] nanoparticles has fuelled significant research in developing strategies for their synthesis and characterization. They are extensively investigated by the chemical route; on the other hand, use of microbial sources for biosynthesis witnessed
Chong Hyun Chang et al.
Nanoscale, 5(4), 1616-1623 (2013-01-22)
A facile electrodeposition technique was utilized to deposit single-walled carbon nanotubes (SWNTs) with cadmium telluride (CdTe) with well-controlled size, density, surface morphology, and composition. By controlling the applied charge, the morphology of these hybrid nanostructures was altered from CdTe nanoparticles
Juan P Monrás et al.
PloS one, 7(11), e48657-e48657 (2012-11-28)
The vast application of fluorescent semiconductor nanoparticles (NPs) or quantum dots (QDs) has prompted the development of new, cheap and safer methods that allow generating QDs with improved biocompatibility. In this context, green or biological QDs production represents a still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service