Skip to Content
Merck
All Photos(2)

Key Documents

145343

Sigma-Aldrich

3,4-Dimethoxycinnamonitrile, predominantly trans

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH=CHCN
CAS Number:
Molecular Weight:
189.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

SMILES string

COc1ccc(\C=C\C#N)cc1OC

InChI

1S/C11H11NO2/c1-13-10-6-5-9(4-3-7-12)8-11(10)14-2/h3-6,8H,1-2H3/b4-3+

InChI key

WGPTVEZJYRFEGT-ONEGZZNKSA-N

General description

3,4-Dimethoxycinnamonitrile undergoes condensation with ethyl glycollate to yield 2-(3,4-dimethoxyphenyl)-3-cyanotetrahydrofuran-4-one[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lignans and related phenols. Part VI. Synthesis of 2-aryltertrahydrooxofuran derivatives.
Ayres DC and Mhasalkar SE.
J. Chem. Soc. Sect. C, 1885-1887 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service