Skip to Content
Merck
All Photos(1)

Key Documents

109797

Sigma-Aldrich

Propionic acid

99%

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
Beilstein:
506071
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021305
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.55 (vs air)

Quality Level

vapor pressure

2.4 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

955 °F

expl. lim.

12.1 %

refractive index

n20/D 1.386 (lit.)

bp

141 °C (lit.)
141 °C

mp

−24-−23 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sa'ad H Al-Lahham et al.
Biochimica et biophysica acta, 1801(11), 1175-1183 (2010-08-10)
Undigested food is fermented in the colon by the microbiota and gives rise to various microbial metabolites. Short-chain fatty acids (SCFA), including acetic, propionic and butyric acid, are the principal metabolites produced. However, most of the literature focuses on butyrate
Weiqiang Chen et al.
Journal of virology, 89(1), 581-593 (2014-10-24)
The recent global resurgence of arthritogenic alphaviruses, in particular chikungunya virus (CHIKV), highlights an urgent need for the development of therapeutic intervention strategies. While there has been significant progress in defining the pathophysiology of alphaviral disease, relatively little is known
R De Kanter et al.
Xenobiotica; the fate of foreign compounds in biological systems, 32(5), 349-362 (2002-06-18)
1. Organ-specific biotransformation was studied in human and rat liver, lung, kidney and small intestine slices and compared on a protein basis, using four model substances. 2. Deethylation of lidocaine was highest in liver slices from both man and rat
B P Espósito et al.
Journal of inorganic biochemistry, 75(1), 55-61 (1999-07-14)
Various divalent rhodium complexes Rh2(L)4 (L = acetate, propionate, butyrate, trifluoroacetate and trifluoroacetamidate) have been found to bind to non-defatted human serum albumin (HSA) at molar ratios about 8:1. The circular dichroism measurements showed that the more liposoluble carboxylates, butyrate
L B Bindels et al.
British journal of cancer, 107(8), 1337-1344 (2012-09-15)
Metabolites released by the gut microbiota may influence host metabolism and immunity. We have tested the hypothesis that inulin-type fructans (ITF), by promoting microbial production of short-chain fatty acids (SCFA), influence cancer cell proliferation outside the gut. Mice transplanted with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service