Merck
  • Formation and occurrence of new polar iodinated disinfection byproducts in drinking water.
Formation and occurrence of new polar iodinated disinfection byproducts in drinking water.
Chemosphere (2015-11-26)
Yang Pan, Wenbin Li, Hao An, Hao Cui, Ying Wang
ABSTRACT
During drinking water disinfection, iodinated disinfection byproducts (I-DBPs) can be generated through reactions between iodide, disinfectants, and natural organic matter. Drinking water I-DBPs have been increasingly attracting attention as emerging organic pollutants as a result of their significantly higher toxicity and growth inhibition than their chloro- and bromo-analogues. In this study, by adopting ultra performance liquid chromatography/electrospray ionization-triple quadrupole mass spectrometry precursor ion scan, multiple reaction monitoring, and product ion scan analyses, 11 new polar I-DBPs with confirmed structures and eight new polar I-DBPs with proposed structures were detected in simulated drinking water samples. Chloramination of simulated raw waters containing natural organic matter with higher aromaticity produced higher levels of new phenolic I-DBPs. Formation of new polar I-DBPs and total organic iodine (TOI) was most favored in chloramination, followed by chlorine dioxide treatment, and relatively minor in chlorination. Lower pH in chloramination substantially enhanced the formation of new polar I-DBPs and TOI. NH2Cl and dissolved organic nitrogen could be important nitrogen sources and precursors for formation of the two new nitrogenous phenolic I-DBPs. Notably, in tap water samples collected from nine major cities located in the Yangtze River Delta region of China, seven of the 11 new polar I-DBPs with confirmed structures were detected at levels from 0.11 to 28 ng/L, and the two new nitrogenous phenolic I-DBPs were ubiquitous with concentrations from 0.12 to 24 ng/L, likely due to the relatively high dissolved organic nitrogen levels in regional source waters.
Sigma-Aldrich
3,5-Diiodosalicylaldehyde, 97%
Sigma-Aldrich
4-Hydroxy-3-methylbenzoic acid, 97%
Sigma-Aldrich
3,5-Diiodosalicylic acid, 99%