Merck
Poly(ethylene glycol)-containing hydrogels in drug delivery.
Journal of controlled release : official journal of the Controlled Release Society (1999-10-16)
N A Peppas, K B Keys, M Torres-Lugo, A M Lowman
ABSTRACT
The use of hydrogels as carriers for protein delivery has been a subject of significant recent research. In our recent work, we have shown that diffusion controlled delivery of proteins from hydrogels containing poly(ethylene glycol) (PEG) can be possible and controlled by the three-dimensional structure. In addition, a number of these hydrogel carriers are mucoadhesive and can be used for protein delivery. PEG star polymer gels have also been prepared by gamma-irradiation and have been used for protein delivery with and without molecular imprinting. The presence of a large number of functional groups in a small volume makes these polymers important for use in biological and pharmaceutical applications. PEG star polymer hydrogels were synthesized using gamma-irradiation and were characterized using swelling techniques. Equilibrium swelling studies were conducted to investigate the effects of molecular weight, number of star arms, concentration, and radiation dose.
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 4,000
Pricing and availability is not currently available.