Skip to Content
Merck
Triethylamine ZerO2®, ≥99%

Triethylamine

Triethylamine ZerO2®, ≥99%
Synonym(s):
Triethylamine, N,N-Diethylethanamine
Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique