Skip to Content
Merck
All Photos(2)

Key Documents

V900335

Sigma-Aldrich

Iodoacetamide

Vetec, reagent grade, 98%

Synonym(s):

2-Iodoacetamide, Monoiodoacetamide, alpha-Iodoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CONH2
CAS Number:
Molecular Weight:
184.96
Beilstein:
1739080
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

92-95 °C (lit.)

storage temp.

2-8°C

SMILES string

NC(=O)CI

InChI

1S/C2H4INO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

PGLTVOMIXTUURA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Iodoacetamide acts as an alkylating reagent for cysteine residues in peptide sequencing. It is an irreversible inhibitor of enzymes with cysteine at the active site. It reacts much more slowly with histidine residues, but that activity is responsible for inhibiting ribonuclease.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirko Zaffagnini et al.
Antioxidants & redox signaling, 16(1), 17-32 (2011-06-29)
Cysteines (Cys) made acidic by the protein environment are generally sensitive to pro-oxidant molecules. Glutathionylation is a post-translational modification that can occur by spontaneous reaction of reduced glutathione (GSH) with oxidized Cys as sulfenic acids (-SOH). The reverse reaction (deglutathionylation)
Valentina R Minciacchi et al.
Oncotarget, 6(13), 11327-11341 (2015-04-11)
Large oncosomes (LO) are atypically large (1-10 µm diameter) cancer-derived extracellular vesicles (EVs), originating from the shedding of membrane blebs and associated with advanced disease. We report that 25% of the proteins, identified by a quantitative proteomics analysis, are differentially
Mia Jüllig et al.
PloS one, 9(5), e96489-e96489 (2014-05-08)
Bypass of foregut secreted factors promoting insulin resistance is hypothesized to be one of the mechanisms by which resolution of type 2 diabetes (T2D) follows roux-en-y gastric bypass (GBP) surgery. To identify insulin resistance-associated proteins and metabolites which decrease more
Jeremy Rouillon et al.
Neuromuscular disorders : NMD, 24(7), 563-573 (2014-05-13)
Diagnosis of muscular dystrophies is currently based on invasive methods requiring muscle biopsies or blood tests. The aim of the present study was to identify urinary biomarkers as a diagnostic tool for muscular dystrophies. Here, the urinary proteomes of Duchenne
Elena B Lugli et al.
Arthritis research & therapy, 17, 9-9 (2015-01-21)
Smoking is a well-established risk factor for rheumatoid arthritis (RA), and it has been proposed that smoking-induced citrullination renders autoantigens immunogenic. To investigate this mechanism, we examined human lung tissue from 40 subjects with defined smoking status, with or without

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service