Skip to Content
Merck
All Photos(1)

Key Documents

1544600

USP

Piroxicam Related Compound J

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 4-hydroxy-2-methyl-2H-benzothiazine-3-carboxylate 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO5S
CAS Number:
Molecular Weight:
269.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

piroxicam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C11H11NO5S/c1-12-9(11(14)17-2)10(13)7-5-3-4-6-8(7)18(12,15)16/h3-6,13H,1-2H3

InChI key

NGHIOTWSWSQQNT-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Piroxicam Related Compound J USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Piroxicam

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service