Skip to Content
Merck
All Photos(1)

Key Documents

52732-U

Supelco

Multi-Layer Silica Gel SPE Tube

configured for US EPA Method 8290, PTFE frit, glass hardware, volume 6 mL, pk of 30

Synonym(s):

Glass Silica Gel SPE (for EPA 8290), 6 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PTFE frit
glass hardware

Quality Level

composition

bed A, 0.1 g (silica)
bed B, 0.2 g (basic silica (2% KOH coated))
bed C, 0.4 g (acidic silica (22% H2SO4 coated))
bed D, 0.2 g (silica)

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

application(s)

food and beverages

compatibility

configured for US EPA Method 8290

General description

Developed specifically for use with US EPA Method 8290 - "Polychlorinated Dibenzodioxins (PCDDs) AND Polychlorinated Dibenzofurans (PCDFs) by High-Resolution Gas Chromatography/High-Resolution Mass Spectrometry (HRGC/HRMS)"

The sample clean-up employed in EPA Method 8290 requires a series of hand-packed glass chromatography steps involving: 1) a mult-layer silica gel class column, 2) a sodium sulfate/alumina glass column, and 3) a multi-layer celite 545-activated carbon glass column.

Cat. No. 52732-U can be used in place of the required multi-layer silica gel glass column, and Cat. No. 52733-U can be used in place of the required multi-layer celite 545-activated carbon glass column.

Note that the bed weights packed into these SPE tubes are smaller than what is described in EPA Method 8290. Therefore, to use these SPE tubes, sample volumes need to be scaled down accordingly.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service