27092-U
Screw cap, solid top with aluminum liner, pkg 100
black phenolic solid cap, aluminum liner, for use with 2 mL vial (standard opening), thread for 8-425
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
aluminum liner
black phenolic solid cap
feature
closure type black phenolic solid top cap
packaging
pkg of 100 ea
parameter
135 °C temp. limit
fitting
thread for 8-425
compatibility
for use with 2 mL vial (standard opening)
Looking for similar products? Visit Product Comparison Guide
General description
Screw caps [for use with 2 mL vial (standard opening), thread 8-425] are available for use with sealing septa and solid caps with aluminum liners. Caps are offered in solid phenolic caps that can withstand a maximum temperature of 135° C.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service