Skip to Content
Merck
All Photos(1)

Key Documents

Z709344

ART® self-sealing barrier pipette tips

ART 5000, volume range 1000-5000 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
maximum volume:
5 mL
volume range:
1000-5000 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 250 ea
manufacturer/tradename:
Molecular BioProducts 2180B

material

colorless

sterility

sterile

feature

barrier

packaging

pack of 250 ea

manufacturer/tradename

Molecular BioProducts 2180B

technique(s)

PCR: suitable

maximum volume

5 mL

volume range

1000-5000 μL

Looking for similar products? Visit Product Comparison Guide

General description

Handle up to 5 mL with this large volume tip

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Gilson Pipetman P-5000 1-5 ml
Nichiryo BenchMate 1-5 ml
Rainin Pipetman P5000 1-5 ml

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service