Skip to Content
Merck
All Photos(1)

Key Documents

SRP6410

Sigma-Aldrich

ApoA-1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Apolipoprotein A-I, MGC117399

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 29 kDa
observed mol wt 26-30 kDa (DTT-reduced. Asp 25 is the predicted N-terminal.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ApoA1(335)

General description

ApoA1 is also known as apolipoprotein A-I, ApoA-I , and is the major protein component of high density lipoprotein (HDL) in plasma. It has a specific role in lipid metabolism. Chylomicrons secreted from the intestinal enterocyte also contain ApoA1 but it is quickly transferred to HDL in the bloodstream. The protein promotes cholesterol efflux from tissues to the liver for excretion. It is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. ApoA-I was also isolated as a prostacyclin (PGI2) stabilizing factor, and thus may have an anticlotting effect. Defects in the gene encoding it are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis. In addition, it has been shown that ApoA1 is implicated in the anti-endotoxin function of HDL via interaction with lipopolysaccharide or endotoxin.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia M Ravodina et al.
ACS omega, 5(36), 23289-23298 (2020-09-22)
Here, we report a nanoparticle-based probe that affords facile cell labeling with cholesterol in cholesterol efflux (CE) assays. This probe, called ezFlux, was optimized through a screening of multiple nanoformulations engineered with a Förster resonance energy transfer (FRET) reporter. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service