Skip to Content
Merck
All Photos(1)

Key Documents

SML3820

Sigma-Aldrich

WH-9943-103C

≥98% (HPLC)

Synonym(s):

N-((1-(2-chloroacetyl)azetidin-3-yl)methyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15ClN2O2
Molecular Weight:
266.72
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

WH-9943-103C, and azetidyl chloroacetamide, is a cell penetrant, selective and potent covalent inhibitor of deubiquitinating enzymes (DUB) VCPIP1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Accelerating inhibitor discovery for deubiquitinating enzymes.
    Nature Communications, 14(1), 686-686 (2023)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service