Skip to Content
Merck
All Photos(1)

Key Documents

SML1962

Sigma-Aldrich

IWP-O1

≥98% (HPLC)

Synonym(s):

IWP-O1, 5-Phenyl-N-(5-phenylpyridin-2-yl)-4-(pyridin-4-yl)-1H-1,2,3-triazol-1-acetamide, IWP-01

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N6O
CAS Number:
Molecular Weight:
432.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]3(nnc(c3c5ccccc5)c4ccncc4)CC(=O)Nc1ncc(cc1)c2ccccc2

InChI key

GYYDQHBKIHQHEB-UHFFFAOYSA-N

Biochem/physiol Actions

IWP-O1 is an inhibitor of Porcupine (Porcn), an acyl (palmitoyl) transferase essential for secretion of Wnt proteins and involved in both embryonic development and tumor progression. IWP-O1 has good metabolic stability and is extremely potent, exhibiting an EC50 value of 80 pM in a cultured cell reporter assay of Wnt signaling.
Porcn Inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service