Skip to Content
Merck
All Photos(1)

Key Documents

SML0944

Sigma-Aldrich

NK-252

≥98% (HPLC)

Synonym(s):

N-[5-(2-Furanyl)-1,3,4-oxadiazol-2-yl]-N′-(2-pyridinylmethyl)-urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N5O3
CAS Number:
Molecular Weight:
285.26
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C13H11N5O3/c19-12(15-8-9-4-1-2-6-14-9)16-13-18-17-11(21-13)10-5-3-7-20-10/h1-7H,8H2,(H2,15,16,18,19)

InChI key

FNSCFQXZZNCDAI-UHFFFAOYSA-N

Biochem/physiol Actions

NK-252 is a very potent, specific activator of the transcription factor Nrf2, which activates antioxidant-associated genes. NK-252 induces expression of an antioxidant response element promoter driven reporter gene, and protects Huh-7 liver cells against H202 induced cytotoxicity.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service