Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702945

Sigma-Aldrich

Anti-5-Methylcytosine(5mC),clone 10S5V10, Rabbit Monoclonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
10S5V10, monoclonal
application:
citations:

biological source

rabbit

Quality Level

material

colorless

clone

10S5V10, monoclonal

form

liquid

concentration

1 mg/mL

color

colorless

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

5mC
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

DB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service