Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300247

Sigma-Aldrich

Monoclonal Anti-MMP9 antibody produced in mouse

clone 5G3, ascites fluid

Synonym(s):

CLG4B, GELB, MANDP2, MMP-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (d)
FACS
IF
IHC
WB
clone:
5G3, monoclonal
species reactivity:
human
citations:
2
technique(s):
direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5G3, monoclonal

mol wt

92 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

Related Categories

Immunogen

Purified recombinant fragment of human MMP9 expressed in E.coli.
Mouse monoclonal antibody raised against MMP9

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengjiang Li et al.
Experimental and therapeutic medicine, 24(6), 750-750 (2022-12-24)
Proline- and serine-rich 2 (PROSER2) is encoded by the 47th open reading frame on human chromosome 10. Bioinformatic analysis has shown PROSER2 was significantly correlated with prognostic outcome of osteosarcoma patients. Its role in the progression and metastasis of human
Qi Gao et al.
Oncotarget, 8(55), 93899-93911 (2017-12-08)
Although materials and modern surgical techniques have been developed to suppress postoperative adhesions, adhesion formation can still occur, and thus, a novel effective anti-adhesion drug is greatly needed. In the present study, we explored the efficacy of paeoniflorin treatment against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service