Skip to Content
Merck
All Photos(1)

Key Documents

SAB5200087

Sigma-Aldrich

Anti-Phosphoserine-Peroxidase antibody produced in rabbit

250 μg/mL, whole antiserum

Synonym(s):

Anti-Phosphoserine:HRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

aqueous solution

concentration

250 μg/mL

technique(s)

indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer)

Specificity

Recognizes proteins phosphorylated on serine residues. Does not cross-react with phosphotyrosine.

Immunogen

Phosphoserine conjugated to KLH

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

HRP Stabilizing buffer

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Li et al.
Nature communications, 10(1), 4996-4996 (2019-11-05)
Plants deploy various immune receptors to recognize pathogens and defend themselves. Crosstalk may happen among receptor-mediated signal transduction pathways in the same host during simultaneous infection of different pathogens. However, the related function of the receptor-like kinases (RLKs) in thwarting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service