Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406107

Sigma-Aldrich

Anti-MBD1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CXXC3, PCM1, RFT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
WB
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
indirect immunofluorescence: suitable
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~66.6 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBD1(4152)

General description

DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. Five transcript variants of the MBD1 are generated by alternative splicing resulting in protein isoforms that contain one MBD domain, two to three cysteine-rich (CXXC) domains, and some differences in the COOH terminus. All five transcript variants repress transcription from methylated promoters; in addition, variants with three CXXC domains also repress unmethylated promoter activity. MBD1 and MBD2 map very close to each other on chromosome 18q21. (provided by RefSeq)

Immunogen

MBD1 (NP_056671.2, 1 a.a. ~ 605 a.a) full-length human protein.

Sequence
MAEDWLDCPALGPGWKRREVFRKSGATCGRSDTYYQSPTGDRIRSKVELTRYLGPACDLTLFDFKQGILCYPAPKAHPVAVASKKRKKPSRPAKTRKRQVGPQSGEVRKEAPRDETKADTDTAPASFPAPGCCENCGISFSGDGTQRQRLKTLCKDCRAQRIAFNREQRMFKRVGCGECAACQVTEDCGACSTCLLQLPHDVASGLFCKCERRRCLRIVERSRGCGVCRGCQTQEDCGHCPICLRPPRPGLRRQWKCVQRRCLRGKHARRKGGCDSKMAARRRPGAQPLPPPPPSQSPEPTEPHPRALAPSPPAEFIYYCVDEDELQPYTNRRQNRKCGACAACLRRMDCGRCDFCCDKPKFGGSNQKRQKCRWRQCLQFAMKRLLPSVWSESEDGAGSPPPYRRRKRPSSARRHHLGPTLKPTLATRTAQPDHTQAPTKQEAGGGFVLPPPGTDLVFLREGASSPVQVPGPVAASTEALLQEAQCSGLSWVVALPQVKQEKADTQDEWTPGTAVLTSPVLVPGCPSKAVDPGLPSVKQEPPDPEEDKEENKDDSASKLAPEEEAGGAGTPVITEIFSLGGTRFRDTAVWLPRSKDLKKPGARKQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service