Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0573-02

Sephadex® G-50

DNA grade, Cytiva, pkg of (pack of 100 g)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115722
NACRES:
NA.52

grade

DNA grade

manufacturer/tradename

Cytiva

packaging

pkg of (pack of 100 g)

particle size

50-150 μm (dry)

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sephadex G-50 grade F is a size exclusion chromatography resin designed for desalting and buffer exchange, and for purification of DNA and oligonucleotides greater than 20 bases in length using spin columns.

Sephadex G-50 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex G-50 is a well-established gel filtration resin for desalting and buffer exchange of biomolecules > 30 000 molecular weight, and with a spin protocol can be used for DNA and oligo purification of molecules greater than 20 bases in length.

Features and Benefits

  • For purification of DNA from small molecules by size exclusion.
  • Tested to ensure reproducibly high recovery of DNA.
  • Sephadex® G-50 DNA Grade is used in illustra Autoseq G-50 columns, ProbeQuant G-50 Micro Columns, MicroSpin G-50 Columns, and NICK Columns.
  • Suitable for researchers who prefer to prepare their own columns for purifying nucleic acids.
  • Excellent for rapid purification of newly synthesized oligonucleotides > 10-mers, including oligonucleotide probes, in 100 to 150 µL of deprotection solution using spin-column chromatography.
  • Useful for desalting or buffer exchange of DNA and removal of unincorporated radionucleotides from end-labeled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 µL.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AutoSeq is a trademark of Cytiva
MicroSpin is a trademark of Cytiva
ProbeQuant is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service