Skip to Content
Merck
All Photos(1)

Key Documents

D2191

Sigma-Aldrich

Monoclonal Anti-DC-SIGN1 antibody produced in mouse

clone 120507, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CD209, Anti-Dendritic Cell-Specific ICAM-3 Grabbing Non-Integrin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

120507, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 2.5 μg using 106 cells
immunocytochemistry: 8-25 μg/mL
immunohistochemistry: 8-25 μg/mL using fixed cells and tissue sections
western blot: 1 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD209(30835)

General description

DC-SIGN is a type II protein C-type lectin that binds mannose-rich molecules such as ICAM-3 and ICAM-2 in a calcium dependent manner. DC-SIGN, a 44 kDa molecule, is identical to the HIV-1 binding protein.

Specificity

Recognizes human DC-SIGN on transfected cells and on monocyte derived dentritic cells. The antibody does not react with either parental mouse cells or irrelevant transfectants, such as human DC-SIGN2.

Immunogen

purified from NIH/3T3 cells transfected with recombinant human DC-SIGN.

Application

Monoclonal Anti-DC-SIGN1 antibody produced in mouse is suitable for flow cytometry, immunocytochemistry at a working concentration of 8-25 μg/mL, immunohistochemistry at 8-25 μg/mL using fixed cells and tissue sections and western blotting at 1 μg/mL.

Biochem/physiol Actions

DC-SIGN1 is involved in molecular interactions between dendritic cells and resting T-cells. DC-SIGN interacts with viruses such as X4, R5, X4R5 HIV-1 strains, HIV-2, and SIV. It captures HIV-1 through its interaction with the gp120 envelope glycoprotein and promotes efficient infection of CD4+ T cells that co-express chemokine receptors.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Tirado-González et al.
Placenta, 33(4), 257-263 (2012-01-21)
Normal pregnancy and spontaneous abortion in humans and mice are associated with immune responses. The decidua harbors dendritic cells identifiable in humans by their expression of DC-SIGN. Because dendritic cells are essential for immune response regulation, decidual DC-SIGN+ cells may
S Pöhlmann et al.
Journal of virology, 75(10), 4664-4672 (2001-04-20)
Dendritic cells (DCs) efficiently bind and transmit human immunodeficiency virus (HIV) to cocultured T cells and so may play an important role in HIV transmission. DC-SIGN, a novel C-type lectin that is expressed in DCs, has recently been shown to
B M Curtis et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(17), 8356-8360 (1992-09-01)
The binding of the human immunodeficiency virus (HIV) envelope glycoprotein gp120 to the cell surface receptor CD4 has been considered a primary determinant of viral tropism. A number of cell types, however, can be infected by the virus, or bind
T B Geijtenbeek et al.
Cell, 100(5), 587-597 (2000-03-18)
Dendritic cells (DC) capture microorganisms that enter peripheral mucosal tissues and then migrate to secondary lymphoid organs, where they present these in antigenic form to resting T cells and thus initiate adaptive immune responses. Here, we describe the properties of
T B Geijtenbeek et al.
Cell, 100(5), 575-585 (2000-03-18)
Contact between dendritic cells (DC) and resting T cells is essential to initiate a primary immune response. Here, we demonstrate that ICAM-3 expressed by resting T cells is important in this first contact with DC. We discovered that instead of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service