Skip to Content
Merck
All Photos(1)

Key Documents

CLS4804

Corning® universal fit pipet tips

1-200 μL universal fit stack rack pipet tips, natural, sterile, 10 racks/case, 4800 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

Pricing and availability is not currently available.

material

natural tip
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation

packaging

rack of 480

manufacturer/tradename

Corning 4804

volume range

1-200 μL

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB2701147SAB4700882SAB4700889
biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse

clone

LF5-1D2, monoclonal

clone

polyclonal

clone

NKTA255, monoclonal

clone

HP-3D9, monoclonal

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

General description

Corning® Universal Fit Pipet Tips

Corning Universal Fit ploypropylene pipet tips are designed to provide a reliable fit with all major brand pipettors and have a bevelled orifice to help ensure fluid delivery. All racked tips are certified RNase/DNase free and nonpyrogenic.

  • Reliable fit with all major brand pipettors
  • Bevelled orifice to help ensure fluid delivery
  • Certified RNase/DNase free and nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service