CLS3324
Corning® CellSTACK® accessories
two vented overcaps (orange) and one solid overcap (red) for the universal cap
Synonym(s):
cell culture chamber accessory
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polyethylene
screw cap
sterility
sterile
feature
disposable
packaging
case of 100
manufacturer/tradename
Corning 3324
color
orange
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Overcaps made from polyethylene
- Disposable screw cap
- Filter adapter overcaps (orange) have peel-away covers for inserting barbed disc filters for sterile venting
- Solid overcap (red) is used in closed systems or applications where venting is not necessary
- Sterlized by gamma irradiation
Available for filling, venting, stacking and manipulation.
Legal Information
CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service