Skip to Content
Merck
All Photos(1)

Key Documents

HL

96121720, human lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human lung
growth mode:
Adherent
karyotype:
Not specified
morphology:
Fibroblast-like
products:
Not specified
receptors:
Not specified

Product Name

HL, 96121720

biological source

human lung

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung fibroblast

Cell Line Description

The cell line HL has been established from normal human lung tissue.

Application

Senescence studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 8,10
D16S539: 12
D5S818: 12
D7S820: 10,11
THO1: 7,9.3
TPOX: 8
vWA: 14,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA;5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service