Skip to Content
Merck
All Photos(1)

Key Documents

EPC

93120820, fish skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

EPC, 93120820, from fish skin

biological source

fish skin

packaging

tube of 5 μg 93120820-DNA-5UG
pkg of vial of cells 93120820-1VL

growth mode

Adherent

karyotype

2n = 96

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

Fish epithelioma Fathead minnow Pimephales promelas

Cell Line Description

This cell line known as Epithelioma papulosum cyprini (EPC) was originally reported to be from Carp (Cyprinus carpio) epidermal herpes virus-induced hyperplastic lesions. More recently EPC was found to be derived from Fathead Minnow (Pimephales promelas) see Winton et al., 2010. This has been confirmed by DNA sequencing at ECACC. The cells support the growth of pancreatic necrosis virus and rhabdovirus.

DNA Profile

Not specified

Culture Medium

EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin/EDTA; 5% CO2; 20 °C. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 and culture for at least one week with 1-2 media changes before shipping.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service